Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB Ligand Rat anti-Mouse, Unlabeled, Clone: TKS-1, BD

Catalog No. BDB550532 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550532 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550532 Supplier BD Biosciences Supplier No. 550532

Rat Monoclonal Antibody

The TKS-1 antibody reacts with 4-1BB Ligand (4-1BBL), a 97kDa member of the NGF/TNF superfamily which is present on antigen-presenting cells and activated B lymphocytes. 4-1BBL interacts with 4-1BB (CDw137), found predominantly on activated T lymphocytes. This ligand-receptor pair is grouped with pairs such as CD27/CD70, CD30/CD153, OX-40/OX-40L, CD80/CD28, and CD86/CD28, which costimulate T-cell proliferation and cytokine production. The interaction of 4-1BB with 4-1BBL may be reciprocally costimulatory in that 4-1BBL may deliver proliferative signals to B lymphocytes activated by anti-IgM. The TKS-1 mAb stains B cells activated for three days with anti-IgM polyclonal antibody, consistent with an earlier report that soluble 4-1BB-alkaline phosphatase fusion protein binds to anti-IgM-activated B cells. Furthermore, 4-1BBL mRNA has been detected in activated T lymphocytes and activated macrophages. The TKS-1 mAb has been reported to inhibit the binding of soluble 4-1BB to 4-1BBL transfectants and block the in vitro costimulation of T-lymphocyte proliferation by 4-1BBL transfectants.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB Ligand
Applications Flow Cytometry
Classification Monoclonal
Clone TKS-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse B lymphoma line 2PK-3
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.