Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4.1N Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB611836 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611836 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611836 Supplier BD Biosciences Supplier No. 611836

Mouse Monoclonal Antibody

The erythrocyte membrane cytoskeletal protein, 4.1R, is an important structural element of red blood cells. It contains a spectrin-actin-binding domain (SABD) that facilitates spectrin interaction with F-actin and a membrane-binding domain (MBD) that links the cytoskeleton to the plasma membrane via interactions with Band 3 protein. Two homologues of 4.1R, 4.1G and 4.1N, are expressed throughout the body and in neurons, respectively. These 4.1 proteins contain Ca2+-dependent (CD-CaM) and Ca2+-independent calmodulin (CI-CaM) binding sites within the MBD domain, a centrally located SABD, and a conserved C-terminal domain (CTD). 4.1N expression is high throughout the adult brain and lower in heart, kidney, and pancreas. In neurons, 4.1N protein co-localizes with PSD-95 and GluR1 at sites of synaptic contacts and the CTD of 4.1N allows binding to the C-terminus of GluR1. In addition, 4.1N interacts with the PI3-Kinase enhancer (PIKE) protein and translocates to the nucleus along with PIKE in response to NGF treatment. Thus, 4.1N is a neuronal membrane cytoskeletal protein that may be involved with localizaton of proteins such as neurotransmitter receptors and signaling molecules. Predominant isoforms of 4.1 proteins have been reported to be observable between 30-210 kD due to complex alternative splicing events. 4.1N has been reported to be observable in a range of 100-135 kD in neuronal populations.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 4.1N
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse 4.1N aa. 510-626
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.