Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4EBP1 (pT36/pT45) Mouse, Alexa Fluor 647, Clone: M31-16, BD

Catalog No. BDB560286 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560286 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560286 Supplier BD Biosciences Supplier No. 560286
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eukaryotic translation initiation factor 4E-Binding Protein hosphorylated heat- and acid-stable protein (PHAS-I or PHAS-1), and it is regulated by insulin. It is a member of the eIF4E-Binding Protein Family, which also includes the proteins 4EBP2 and 4EBP3. 4EBP1 binds with eukaryotic translation Initiation Factor 4E (eIF4E), which prevents its assembly into the eIF4E complex and inhibits cap-dependent translation. When 4EBP1 is phosphorylated, this binding is disrupted, allowing cap-dependent translation to be activated. Phosphorylation of 4EBP1 is required for protein synthesis, and it mediates the regulation of protein translation by stimuli that signal through the phosphoinositide 3 (PI3) kinase pathway. We found that threonines 36 and 45 (T36/T45) are phosphorylated in resting human peripheral blood mononuclear cells. PI3 kinase inhibitors, such as LY294002 down-regulate the phosphorylation level of 4EBP1 (pT36/pT45).

The M31-16 monoclonal antibody recognizes the phosphorylated T36 and T45 of activated human 4EBP1. The orthologous phosphorylation sites in mouse and rat 4EBP1 are T35 and T44.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 4EBP1 (pT36/pT45)
Applications Flow Cytometry
Classification Monoclonal
Clone M31-16
Conjugate Alexa Fluor 647
Description 4E-BP1. EIF4EBP1, P/OKCL.6, PHAS-I, PHAS-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human 4EBP1 (pT45) Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.