Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4F2 hc/CD98HC Mouse anti-Rat, Unlabeled, Clone: 30, BD

Catalog No. BDB611516 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611516 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611516 Supplier BD Biosciences Supplier No. 611516
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

4F2 antigen (CD98) was identified as a heterodimer consisting of an 80 kDa type II glycosylated integral membrane protein 4F2 heavy chain (4F2 hc) linked by disulfide bonds to a hydrophobic, non-glycosylated, 37 kDa protein 4F2 light chain (4F2 lc). The 4F2 hc colocalizes with cadherins at cell adhesion sites and is required for the intracellular targeting of 4F2 heterodimers. 4F2 hc also associates with β1 integrins and overexpression of 4F2 reverses the suppression of β1 integrin activation caused by overexpression of β1 cytoplasmic domains. 4F2 has also been identified as a fusion regulatory protein FRP-1, since anti-FRP-1 antibodies can induce HIV-mediated cell fusion via an integrin system. 4F2 heterodimers are also important for activation of amino acid transport. 4F2 hc is 30% homologous with the amino acid transport activator D2/rBAT and expression of 4F2 hc in Xenopus oocytes induces system y+L amino acid transport. Thus, 4F2 hc may participate in intracellular trafficking and activation of amino acid transporters, as well as in the regulation of integrin signaling.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 4F2 hc/CD98HC
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Description 4F2 hc
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat 4F2 hc aa. 9-204
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.