Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxymethylcytosine (5hmC) Mouse anti-All, Clone: HMC-MA01, MilliporeSigma™

Catalog No. MABE317MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE317MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE317MI Supplier MilliporeSigma Supplier No. MABE317
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect 5-Hydroxymethylcytosine (5hmC) using this Anti-5-Hydroxymethylcytosine (5hmC) Antibody, clone HMC-MA01 validated for use in Dot Blot, immunocytochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-Hydroxymethylcytosine (5hmC)
Applications Dot Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone HMC-MA01
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen BSA-conjugated recombinant Hydroxymethylcytosine DNA.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.