Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-methylcytosine Mouse anti-Human, Mouse, Clone: 33D3, MilliporeSigma™

Catalog No. MABE146MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE146MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE146MI Supplier MilliporeSigma Supplier No. MABE146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-5-methylcytosine Antibody, clone 33D3 (mouse monoclonal antibody) validated in DB, immunofluoresence, SB, immunohistochemistry (paraffin), Flow Cytometry, MeDimmunoprecipitation to detect 5-methylcytosine also known as methylcytosine, methylcytidine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-methylcytosine
Applications ChIP Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 33D3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1λ in buffer containing PBS without preservative.
Host Species Mouse
Immunogen Ovalbumin-conjugated 5-methylcytosine
Purification Method Protein A
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.