Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
53BP1 Mouse anti-Human, Clone: BP13, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05726
Description
Detect 53BP1 with Anti-53BP1 Antibody, clone BP13 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
| 53BP1 | |
| Monoclonal | |
| Unconjugated | |
| NM_005657.1 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C for 24 months. | |
| IgG1 |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| BP13 | |
| 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
| TP53BP1, 53BP1, p53BP1, p202, FLJ41424, MGC138366 | |
| Mix of three GST fusion proteins corresponding to residues 1-337, 338-671, and 1331-1664, respectively, of human 53BP1 | |
| 200 μg | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction