Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Mouse anti-Human, Clone: BP13, MilliporeSigma™

Catalog No. MAB3802MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3802MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3802MI Supplier MilliporeSigma Supplier No. MAB3802
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect 53BP1 also known as p53 Binding Protein 1 with Anti-53BP1 Antibody, clone BP13 (Mouse Monoclonal Antibody) validated in IF and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone BP13
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS, containing sodium azide.
Gene Accession No. NM_005657.1
Gene Symbols TP53BP1, 53BP1, MGC138366, p202, p53BP1, FLJ41424
Host Species Mouse
Immunogen Recombinant human 53BP1 proteins.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.