Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Mouse anti-Human, Mouse, Clone: BP18, MilliporeSigma™

Catalog No. 05725MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-725-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-725-MI Supplier MilliporeSigma Supplier No. 05725
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect 53BP1 with Anti-53BP1 Antibody, clone BP18 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BP18
Conjugate Unconjugated
Formulation mouse ascites IgM containing 0.05% sodium azide and 30% glycerol
Gene Accession No. NM_005657.1
Gene Symbols TP53BP1, 53BP1, p53BP1, p202, FLJ41424, MGC138366
Host Species Mouse
Immunogen Mix of three GST fusion proteins corresponding to residues 1-337, 338-671, and 1331-1664, respectively, of human 53BP1
Purification Method Ascites
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.