Learn More
Invitrogen™ 53BP1 Polyclonal Antibody, DyLight™ 488

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522759
Description
Human samples have been tested in Western Blot and ICC/IF, and mouse samples have been tested in ICC/IF only. Suggested positive control: U205, 293T, and MO59K cell lysates and MEF lysates. Predicted cross-reactivity based on sequence identity: Chimpazee/Equine/Feline/Gibbon/Gorilla/Hamster/Marmoset/Orangutan/Panda/Porcine/Rabbit/Sheep (100%).
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Specifications
| 53BP1 | |
| Polyclonal | |
| DyLight 488 | |
| TP53BP1 | |
| 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27223, 296099, 478274, 509111, 7158 | |
| 4°C, store in dark | |
| Liquid |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| P70399, Q12888 | |
| TP53BP1, Trp53bp1 | |
| Region between residues 1925 and the C-terminus (residue 1972) of human 53BP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Bovine, Rodent, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.