Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6xHis Mouse, Unlabeled, Clone: F24-796, BD

Catalog No. BDB552565 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552565 Supplier BD Biosciences Supplier No. 552565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The engineering of a short epitope tag to a recombinant protein provides a very convenient method for detecting and isolating the protein of interest. For example, the placement of a series of 6 consecutive histidine residues that are incorporated at either end of the recombinant fusion protein facilitates the purification of that protein. Purification can be easily accomplished by metal chelate affinity chromatography using a metal resin column. The 6xHis monoclonal antibody can be used to detect the epitope-tagged protein of interest.

TRUSTED_SUSTAINABILITY

Specifications

Antigen 6xHis
Applications Western Blot
Classification Monoclonal
Clone F24-796
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.