Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxydeoxyguanosine Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB5830MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5830MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5830MI Supplier MilliporeSigma Supplier No. AB5830
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This Anti-8-Hydroxydeoxyguanosine Antibody is validated for use in ELISA, immunohistochemistry (paraffin) for the detection of 8-Hydroxydeoxyguanosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 8-Hydroxydeoxyguanosine
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Goat Serum. Liquid containing 0.01% Thimerosal.
Host Species Goat
Immunogen 8-Hydroxy-2'-deoxyguanosine and 8-hydroxyguanosine.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.