Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-8-Hydroxyguanosine, Polyclonal

Catalog No. DR1001100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR1001100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR1001100UL Supplier MilliporeSigma™ Supplier No. DR1001100UL
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects 8-Hydroxyguanosine Clone: in samples, and it is validated for Immunohistochemistry (Paraffin)

8-Hydroxydeoxy-guanosine is a modified base that occurs in DNA due to attack by reactive oxygen species. Increases in the levels of 8-hydroxyguanosine have been associated with several diseases in humans, such as arteriosclerosis, cerebral and heart ischemia-reperfusion injury, cancer, rheumatoid arthritis, inflammation, diabetes, aging, and neurodegenerative conditions, such as Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 8-Hydroxyguanosine
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Goat
Immunogen 8-Hydroxyguanosine
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes 8-Hydroxydeoxyguanosine in Alzheimer's disease brain tissue.
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.