Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Oxoguanine Mouse anti-Human, Rat, Mouse, Monkey, Clone: 483.15, MilliporeSigma™

Catalog No. MAB3560MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3560MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3560MI Supplier MilliporeSigma Supplier No. MAB3560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-8-Oxoguanine Antibody, clone 483.15 detects level of 8-Oxoguanine and has been published and validated for use in ELISA and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 8-Oxoguanine
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 483.15
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal IgM liquid in buffer containing no preservative.
Host Species Mouse
Immunogen 8-oxoguanine adsorbed on to alumina
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.