Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A-Raf Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610074 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610074 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610074 Supplier BD Biosciences Supplier No. 610074
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A-Raf, a proto-oncogene that encodes a 68 kDa serine/threonine-specific protein kinase, shows strong homology to c-Raf and v-Raf in the kinase region of the molecule. c-Raf appears to be uniformly and ubiquitously expressed, but A-Raf expression is much more variable. A-Raf is found at highest levels in urogenital and kidney tissues, with the lowest levels found in brain. Cell transformation by A-Raf has been shown to be independent of p21[ras], suggesting the possibility of an independent signal transduction pathway.

TRUSTED_SUSTAINABILITY

Specifications

Antigen A-Raf
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse A-Raf aa. 183-199
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.