Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-A20 Clone: 59A426,

Catalog No. AM63100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AM63100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM63100UG Supplier MilliporeSigma™ Supplier No. AM63100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects A20 Clone: 59A426 in Human samples, and it is validated for Immunoprecipitation

NF-κB (nuclear factor κB) regulates the expression of a large number of genes that play critical roles in apoptosis, viral replication, tumorigenesis, various autoimmune diseases and inflammation. The zinc finger protein A20 is a tumor necrosis factor and interleukin 1 (IL-1)-inducible protein that negatively regulates NF-κB gene expression by interferring with a novel TNF-induced and RIP- or TRAF2-mediated pathway that is different from the NIK-IκB kinase pathway. A20, an 70kDa protein, is expressed at low levels in most organs, the highest expression levels being in lymphoid tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A20
Applications Immunoprecipitation
Classification Monoclonal
Clone 59A426
Conjugate Unconjugated
Formulation In PBS, 0.2% gelatin.
Host Species Mouse
Immunogen full-length recombinant human A20 protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽70kDa A20 protein in Daudi cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.