Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2B5 Mouse, Alexa Fluor 647, Clone: 105/A2B5, BD

Catalog No. BDB563776 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563776 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563776 Supplier BD Biosciences Supplier No. 563776
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 105/A2B5 monoclonal antibody binds to gangliosides in neural tissue in the retina, brain, spinal cord, and dorsal root ganglia. The antigenic gangliosides are found on the plasma membrane of neural cell bodies, but not axons or dendrites. Expression levels of the gangliosides that mAb 105/A2B5 binds to, including GT3 and its O-acetylated derivative, decrease during embryonic development of the brain. Thus, mAb 105/A2B5 may be used to monitor the maturation of brain tissue. A2B5 antigens have also been associated with numerous tumors, including glioblastoma multiforme tumor cells, pancreatic islet tumor cells, and rat insulinoma cells, and are thought to contribute to chemoresistance and increased tumor proliferation and recurrence.

TRUSTED_SUSTAINABILITY

Specifications

Antigen A2B5
Applications Flow Cytometry
Classification Monoclonal
Clone 105/A2B5
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Chicken embryo retina Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.