Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2B5 Mouse anti-Mammals, Clone: A2B5-105, MilliporeSigma™

Catalog No. MAB312RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB312RMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB312RMI Supplier MilliporeSigma Supplier No. MAB312R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-A2B5 Antibody, clone A2B5-105 detects level of A2B5 and has been published and validated for use in flow cytometry, immunocytochemistry, immunofluoresence, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2B5
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone A2B5-105
Conjugate Unconjugated
Formulation Liquid. Buffer = PBS, pH 7.1 with 2mg/mL BSA.
Gene Symbols A2B5
Host Species Mouse
Immunogen Embryonic chicken retinal cells
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.