Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-A2B5, Clone: A2B5-105, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB312R/DEL
Description
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry

Specifications
A2B5 | |
Unconjugated | |
Embryonic chicken retinal cells | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Neuroscience | |
Mammalia | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction