Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-A2M, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Supplier: Abnova Corporation H00000002PW1
Includes: Antibody pair for IP: rabbit polyclonal anti-A2M (300μL) 2.Antibody pair for WB: mouse purified polyclonal anti-A2M (50μg)
Specifications
A2M | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
CPAMD5/DKFZp779B086/FWP007/S863-7 | |
Mouse, Rabbit | |
RUO | |
2 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
A2M | |
A2M | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-A2M (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-A2M (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction