Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

apoptosis antagonizing transcription factor, Mouse, Clone: 3C7, Abnova™

Catalog No. 89001246 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-246 Supplier Abnova Corporation Supplier No. H00026574M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AATF.

The protein encoded by this gene was identified on the basis of its interaction with MAP3K12/DLK, a protein kinase known to be involved in the induction of cell apoptosis. This gene product contains a leucine zipper, which is a characteristic motif of transcription factors, and was shown to exhibit strong transactivation activity when fused to Gal4 DNA binding domain. Overexpression of this gene interfered with MAP3K12 induced apoptosis. [provided by RefSeq

Sequence: VQSISDFEKFTKGMDDLGSSEEEEDEESGMEEGDDAEDSQGESEEDRAGDRNSEDDGVVMTFSSVKVSEEVEKGRAVKNQIALWDQLLEGRIKLQKALLT

Specifications

Antigen apoptosis antagonizing transcription factor
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AATF.
Formulation PBS with no preservative; pH 7.4
Gene AATF
Gene Accession No. BC000591
Gene Alias CHE-1/CHE1/DED
Gene Symbols AATF
Host Species Mouse
Immunogen AATF (AAH00591, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 26574
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.