Learn More
ABCA8, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ABCA8 peptide using ARM Technology.
Supplier: Abnova Corporation H00010351K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. The function of this protein has not yet been determined. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ABCA8 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ABCA8 | |
ABCA8 | |
A synthetic peptide of human ABCA8 | |
RUO | |
10351 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ATP-binding cassette, subfamily A (ABC1), member 8 | |
ABCA8 | |
KIAA0822, MGC163152 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.