Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP-binding cassette, sub-family B (MDR/TAP), member 9, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-563 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-563 Supplier Abnova Corporation Supplier No. H00023457A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ABCB9.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. The function of this half-transporter has not yet been determined; however, this protein may play a role in lysosomes. Alternative splicing of this gene results in distinct isoforms which are likely to have different substrate specifications. [provided by RefSeq

Sequence: FIDRQPTMVHDGSLAPDHLEGRVDFENVTFTYRTRPHTQVLQNVSFSLSPGKVTALVGPSGSGKSSCVNILENFYPLEGGRVLLDGKPISAYDHKYLHR

Specifications

Antigen ATP-binding cassette, sub-family B (MDR/TAP), member 9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ABCB9.
Formulation 50% glycerol
Gene ABCB9
Gene Accession No. NM_019625
Gene Alias EST122234/KIAA1520/TAPL
Gene Symbols ABCB9
Host Species Mouse
Immunogen ABCB9 (NP_062571, 482 a.a. ∼ 580 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23457
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.