Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-189 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-189 Supplier Abnova Corporation Supplier No. H00084696B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ABHD1 protein.

This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions. [provided by RefSeq

Sequence: MLSSFLSPQNGTWADTFSLLLALAVALYLGYYWACVLQRPRLVAGPQFLAFLEPHCSITTETFYPTLWCFEGRLQSIFQVLLQSQPLVLYQSDILQTPDGGQLLLDWAKQPDSSQDPDPTTQPIVLLLPGITGSSQETYVLHLVNQALRDGYQAVVFNNRGCRGEELRTHRAFCASNTEDLETVVNHIKHRYPQAPLLAVGISFGGILVLNHLAQARQAAGLVAALTLSACWDSFETTRSLETPLNSLLFNQPLTAGLCQLVERNRKVIEKVVDIDFVLQARTIRQFDERYTSVAFGYQDCVTYYKAASPRTKIDAIRIPVLYLSAADDPFSPVCALPIQAAQHSPYVALLITARGGHIGFLEGLLPWQHWYMSRLLHQYAKAIFQDPEGLPDLRALLPSEDRNS

Specifications

Antigen ABHD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ABHD1 protein.
Formulation PBS with no preservative; pH 7.4
Gene ABHD1
Gene Accession No. BC039576.1
Gene Alias FLJ36128/LABH1
Gene Symbols ABHD1
Host Species Mouse
Immunogen ABHD1 (AAH39576.2, 1 a.a. ∼ 405 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84696
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.