Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

abhydrolase domain containing 5, Mouse, Clone: 4B12, Abnova™

Catalog No. 89001273 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-273 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-273 Supplier Abnova Corporation Supplier No. H00051099M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ABHD5.

The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation. [provided by RefSeq

Sequence: FEDDTVTEYIYHCNVQTPSGETAFKNMTIPYGWAKRPMLQRIGKMHPDIPVSVIFGARSCIDGNSGTSIQSLRPHSYVKTIAILGAGHYVYADQPEEFNQKVK

Specifications

Antigen abhydrolase domain containing 5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ABHD5.
Formulation PBS with no preservative; pH 7.4
Gene ABHD5
Gene Accession No. NM_016006
Gene Alias CDS/CGI58/IECN2/MGC8731/NCIE2
Gene Symbols ABHD5
Host Species Mouse
Immunogen ABHD5 (NP_057090, 240 a.a. ∼ 342 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51099
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.