Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abl SH3 Domain Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06466MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-466-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-466-MI Supplier MilliporeSigma Supplier No. 06466

Rabbit Polyclonal Antibody

Anti-Abl SH3 Domain Antibody detects level of Abl SH3 Domain and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Abl SH3 Domain
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine buffer, pH 7.4
Gene Symbols ABL1; v-abl; JTK7; p150; c-ABL; bcr/abl; ABL
Host Species Rabbit
Immunogen Recombinant protein containing GST fused to the SH3 domain of human p210bcr-abl corresponding to amino acids 929-1020
Purification Method Protein A chromatography
Quantity 250 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_009594
Target Species Human, Mouse
Content And Storage −20°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.