Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABL2, Mouse, Clone: 6D5, Abnova™

Catalog No. 89016009 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-009 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-009 Supplier Abnova Corporation Supplier No. H00000027M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ABL2.

This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq

Sequence: KKTLGLRAGKPTASDDTSKPFPRSNSTSSMSSGLPEQDRMAMTLPRNCQRSKLQLERTVSTSSQPEENVDRANDMLPKKSEESAAPSRERPKAKLLPRGA

Specifications

Antigen ABL2
Applications ELISA, In situ PLA, KnockDown, Western Blot
Classification Monoclonal
Clone 6D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ABL2.
Formulation PBS with no preservative; pH 7.4
Gene ABL2
Gene Accession No. BC065912
Gene Alias ABLL/ARG/FLJ22224/FLJ31718/FLJ41441
Gene Symbols ABL2
Host Species Mouse
Immunogen ABL2 (AAH65912, 743 a.a. ∼ 842 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.