Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABLIM3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-834 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-834 Supplier Abnova Corporation Supplier No. H00022885B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ABLIM3 protein.

The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 [PubMed 16328021]).[supplied by OMIM

Sequence: MNTSIPYQQNPYNPRGSSNVIQCYRCGDTCKGEVVRVHNNHFHIRCFTCQVCGCGLAQSGFFFKNQEYICTQDYQQLYGTRCDSCRDFITGEVISALGRTYHPKCFVCSLCRKPFPIGDKVTFSGKECVCQTCSQSMASSKPIKIRGPSHCAGCKEEIKHGQSLLALDKQWHVSCFKCQTCSVILTGEYISKDGVPYCESDYHAQFGIKCETCDRYISGRVLEAGGKHYHPTCARCVRCHQMFTEGEEMYLTGSEVWHPICKQAARAEKKLKHRRTSETSISPPGSSIGSPNRVICDIYENLDLRQRRASSPGYIDSPTYSRQGMSPTFSRSPHHYYRSGDLSTATKSKTSEDISQTSKYSPIYSPDPYYASESEYWTYHGSPKVPRARRFSSGGEEDDFDRSMHKLQSGIGRLILKEEMKARSSSYADPWTPPRSSTSSREALHTAGYEMSLNGSPRSHYLADSDPLISKSASLPAYRRNGLHRTPSADLFHYDSMNAVNWGMREYKIYPYELLLVTTRGRNRLPKDVDRTRLEGNFWKSGCL

Specifications

Antigen ABLIM3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ABLIM3 protein.
Formulation PBS with no preservative; pH 7.4
Gene ABLIM3
Gene Accession No. BC001665.2
Gene Alias HMFN1661
Gene Symbols ABLIM3
Host Species Mouse
Immunogen ABLIM3 (AAH01665.1, 1 a.a. ∼ 544 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22885
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.