Learn More
ACADS, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ACADS peptide using ARM Technology.
Supplier:  Abnova Corporation H00000035K
Catalog No. 89-200-290
Request a Quote
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Description
This gene encodes a a tetrameric mitochondrial flavoprotein, which is a member of the acyl-CoA dehydrogenase family. This enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Mutations in this gene have been associated with Short Chain Acyl-CoA Dehydrogenase Deficiency. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line
Order Info
  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
ACADS
Monoclonal
Unconjugated
In 1X PBS, pH 7.4
ACADS
ACADS
A synthetic peptide of human ACADS
RUO
35
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Western Blot
1mg
acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain
ACADS
ACAD3, SCAD
Rabbit
1 mg x up to 3 Abs
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.