Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAP3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525101
Description
ACAP3 is GTPase-activating protein for the ADP ribosylation factor family.
Specifications
ACAP3 | |
Polyclonal | |
ACAP3 | |
CENTB5, arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 3, centaurin, beta 5, centaurin-beta-5, cnt-b5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116983 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q96P50 | |
ACAP3 | |
KLH conjugated synthetic peptide between 101-130 amino acids from the N-terminal region of human ACAP3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction