Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT-2 Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. ABS988MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS988MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS988MI Supplier MilliporeSigma Supplier No. ABS988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-ACAT-2 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of ACAT-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAT-2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit Polyclonal in buffer containing 20mM NaPO4, 28mM NaCl and 0.05% sodium azide.
Gene Symbols SOAT2; ACACT2; ACAT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACAT-2.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003569
Target Species Human, Monkey
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.