Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE, Mouse, Clone: 6A4, Abnova™

Catalog No. 89021272 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-272 Supplier Abnova Corporation Supplier No. H00001636M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ACE.

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined. [provided by RefSeq

Sequence: RLATAMKLGFSRPWPEAMQLITGQPNMSASAMLSYFKPLLDWLRTENELHGEKLGWPQYNWTPNSDDFYNETETKIFLQFYDQTGIWDHGAPHLLPPSQARGTREAPVYM

Specifications

Antigen ACE
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ACE.
Formulation PBS with no preservative; pH 7.4
Gene ACE
Gene Accession No. BC036375
Gene Alias ACE1/CD143/DCP/DCP1/MGC26566
Gene Symbols ACE
Host Species Mouse
Immunogen ACE (AAH36375, 592 a.a. ∼ 701 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1636
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.