Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl CoA Carboxylase Mouse anti-Human, Rat, Clone: 7D2.2, MilliporeSigma™

Catalog No. 051098MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-109-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-109-8MI Supplier MilliporeSigma Supplier No. 051098
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-acetyl CoA Carboxylase Antibody, clone 7D2.2 detects level of acetyl CoA Carboxylase and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl CoA Carboxylase
Applications Western Blot
Classification Monoclonal
Clone 7D2.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_942131
Gene Symbols ACAC, ACC, ACC-alpha, ACC1, ACCA
Host Species Mouse
Immunogen Histidine-tagged recombinant protein corresponding to human acetyl CoA Carboxylase.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.