Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H2A (Lys9) Rabbit anti-Human, Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 07289MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-289-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-289-MI Supplier MilliporeSigma Supplier No. 07289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-Histone H2A (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H2A (Lys9) also known as H2AK9Ac, Histone H2A (acetyl K9) and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H2A (Lys9)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation antiserum immuno-depleted with the non-acetylated immunogenic peptide, containing 0.05% sodium azide and 30% glycerol
Gene Symbols H2AFX; H2A/X; H2A.X; H2AX; H2a/x
Host Species Rabbit
Immunogen Peptide (GKQGGAcKARAKAK-C) corresponding to amino acids 4-15 of human Histone H2A.
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003517
Target Species Human, Vertebrates
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.