Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl Histone H2A.Z (Lys 5, 7, 11) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE1363MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1363MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1363MI Supplier MilliporeSigma Supplier No. ABE1363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-acetyl Histone H2A.Z (Lys 5, 7, 11) Antibody is validated for use in Western Blotting and Immunocytochemistry and Chromatin Immunoprecipitation (ChIP) and ChIP-seq and Dot Blot and ELISA for the detection of acetyl Histone H2A.Z (Lys 5, 7, 11).
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl Histone H2A.Z (Lys 5
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide and 0.05% ProClin300.
Gene Symbols H2AFZ; H2AZ
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human acetyl Histone H2A.Z (Lys 5, 7, 11) near the N-terminus.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_002097
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.