Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H2B (Lys24) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABE269MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE269MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE269MI Supplier MilliporeSigma Supplier No. ABE269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-acetyl-Histone H2B (Lys24) Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation, DB to detect acetyl-Histone H2B (Lys24) also known as H2BK24Ac, Histone H2B (acetyl K24).
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H2B (Lys24)
Applications Dot Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST3H2BB
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H2B acetylated at Lys24.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001019770
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.