Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H2B Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07373MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-373-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-373-MI Supplier MilliporeSigma Supplier No. 07373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect acetyl-Histone H2B also known as Histone H2B with Anti-acetyl-Histone H2B Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H2B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols HIST3H2BB
Host Species Rabbit
Immunogen Synthetic peptide in which there is an acetyl-lysine at residues 5, 12, 15 and 20 of human Histone H2B
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_778225
Target Species Human
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.