Learn More
Acetyl-Histone H3 (Lys 4) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | acetyl-Histone H3 (Lys 4) |
| Applications | Multiplex, ChIP sequencing (ChIP-seq), Dot Blot, Immunoassay, Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 100μl of immunoaffinity purified rabbit IgG in a buffer with 0.1M Tris-glycine, pH 7.4 and 0.15M NaCl, and containing 0.05% sodium azide and 30% glycerol |
| Gene Symbols | H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide containing the sequence ...RT[AcK]Q... in which [AcK] corresponds to acetyl lysine 4 of human histone H3 |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.