Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H3 (Lys14) Mouse anti-Human, Mouse, Clone: 13HH3-1A5, MilliporeSigma™

Catalog No. MABE351MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE351MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE351MI Supplier MilliporeSigma Supplier No. MABE351
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-acetyl-Histone H3 (Lys14) Antibody, clone 13HH3-1A5 is a mouse monoclonal antibody for detection of acetyl-Histone H3 (Lys14) also known as Histone H3.1t, H3/t, H3t, H3/g and has been validated in western blot, immunoprecipitation, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H3 (Lys14)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 13HH3-1A5
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 in ascites containing 0.05% sodium azide.
Gene Accession No. NP_003484
Gene Symbols HIST3H3, H3FT
Host Species Mouse
Immunogen Ovalbumin-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys14.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.