Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Histone H3 (Lys23) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$160.70 - $684.50
Specifications
Antigen | acetyl-Histone H3 (Lys23) |
---|---|
Applications | ChIP Assay, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Serum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
07-355-SMI
|
MilliporeSigma
07355S |
10 μL |
Each for $160.70
|
|
|||||
07-355-MI
|
MilliporeSigma
07355 |
100 μL |
Each for $684.50
|
|
|||||
Description
Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).Specifications
acetyl-Histone H3 (Lys23) | |
Polyclonal | |
Serum | |
Human, S. cerevisiae, Vertebrates, Yeast | |
NM_003493 | |
Ovalbumin-conjugated, synthetic peptide (KQLASAcKAARK-C) corresponding to amino acids 18-27 of yeast histone H3 acetylated on lysine 23 with a C-terminal cysteine added for conjugation purposes | |
Antiserum |
ChIP Assay, Western Blot | |
Unconjugated | |
Rabbit | |
Rabbit antiserum containing 0.05% sodium azide and 30% glycerol | |
HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t | |
Primary | |
−20°C for 24 months |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title