Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-acetyl Histone H3(Lys27), Polyclonal,

Catalog No. ABE1362MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1362MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1362MI Supplier MilliporeSigma™ Supplier No. ABE1362

Specifications

Antigen acetyl Histone H3(Lys27)
Applications ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal in PBS containing 0.05% azide and 0.05% ProClin 300.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human acetyl Histone H3 (Lys27) near the N-terminal.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003520
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.