Learn More
Specifications
Specifications
| Antigen | acetyl-Histone H3 (Lys79) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit IgG in 70% storage buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, 5mg/mL BSA and 30% glycerol. |
| Gene Symbols | H3F3A; H3.3B; H3F3; MGC87783; H3F3B; H3.3A; MGC87782 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated, synthetic peptide containing the sequence (...DF[acK]TD...) where [AcK] corresponds to acetyl-lysine 79 of human histone H3. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.