Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Histone H3 (Lys9/18) Rabbit anti-Bovine, Human, Mouse, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07593
Description
Anti-acetyl-Histone H3 (Lys9/18) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys9/18) also known as H3K9/18Ac, Histone H3 (acetyl K9/18) and has been validated in Chimmunoprecipitation, western blot, Mplex.
Specifications
acetyl-Histone H3 (Lys9/18) | |
Polyclonal | |
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative. | |
Rabbit | |
Unpurified | |
Primary | |
Bovine, Human, Mouse, S. cerevisiae, Yeast | |
Serum |
Multiplex, Immunoassay, Western Blot | |
Unconjugated | |
H3F3A; MGC87783; H3.3A; MGC87782; H3F3; H3.3B; H3F3B | |
peptide (ARAcKSTGGKAPRAcKQL) which corresponds to acetyl-lysine at residue 9 and 18 of human Histone H3 | |
200 μL | |
NM_003493 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction