Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H3 (Lys9) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Catalog No. 07352MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-352-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-352-MI Supplier MilliporeSigma Supplier No. 07352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H3 (Lys9) also known as H3K9Ac, Histone H3 (acetyl K9) and has been published and validated in Chimmunoprecipitation, western blot, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H3 (Lys9)
Applications Multiplex, Immunoassay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Antiserum containing 0.35% sodium azide and 30% glycerol.
Gene Symbols HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Host Species Rabbit
Immunogen Ovalbumin-conjugated, synthetic peptide (KQTARAcKSTGGK-C) corresponding to amino acids 4-14 of yeast histone H3 acetylated on lysine 9, with a C-terminal cysteine added for conjugation purposes.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Human, S. cerevisiae, Vertebrates, Yeast
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.