Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl Histone H3 (Lys9) Rabbit anti-Human, Mouse, Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$144.24 - $615.02
Specifications
Antigen | acetyl Histone H3 (Lys9) |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Target Species | Chicken, Human, Mouse |
Description
Anti-acetyl Histone H3 (Lys9) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated at lysine 9. This purified antibody, also known as Anti-H4K9ac has dot blot (DB) proven specificity and has been validated in western blot, DB, Chimmunoprecipitation, Mplex.Specifications
acetyl Histone H3 (Lys9) | |
Unconjugated | |
Chicken, Human, Mouse | |
NP_003484 | |
KLH-conjugated linear peptide corresponding to human Histone H3 acetylated at Lys9. | |
Affinity Purified |
Polyclonal | |
Rabbit | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 | |
Primary | |
2-8°C for 12 months |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title