Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H4 Rabbit anti-Human, Tetrahymena, Eukaryote, Polyclonal, MilliporeSigma™

Catalog No. 06866MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-866-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-866-MI Supplier MilliporeSigma Supplier No. 06866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in Chimmunoprecipitation, western blot, immunocytochemistry to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H4
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Whole antiserum containing 0.05% sodium azide.
Gene Symbols HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; IST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; 4FJ; H4FD; H4/I; H4/B; H4/D; H4/C; HIST1H4J; HIST1H4D; H4/E
Host Species Rabbit
Immunogen KLH-conjugated peptide corresponding to a.a. 2-19 of Tetrahymena histone H4 [AGGAcKGGAcKGMGAcKVGAAcKRHS-C], acetylated on lysines 5, 8, 12 and 16.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_175054
Target Species Eukaryote, Human, Tetrahymena species
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.