Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Histone H4 (Lys12) Rabbit anti-Human, Vertebrates, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04119
Description
This Anti-acetyl-Histone H4 (Lys12) Antibody, rabbit is validated for use in western blotting for the detection of acetyl-Histone H4 (Lys12).
Specifications
acetyl-Histone H4 (Lys12) | |
Monoclonal | |
100μL of Rabbit Monoclonal IgG cell culture supernatant with 0.1% sodium azide. | |
Rabbit | |
100 μL | |
HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, | |
−20°C for 24 months | |
IgG |
ChIP Assay, Dot Blot, Western Blot | |
Unconjugated | |
NM_003538.3 | |
Peptide corresponding to Histone H4 containing the sequence [GLG-AcK-GGA] on which Lys12 is acetylated. | |
Primary | |
Human, Vertebrates | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction