Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl Histone H4 (Lys12) Rabbit anti-Human, Yeast, Polyclonal, MilliporeSigma™

Catalog No. ABE532MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE532MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE532MI Supplier MilliporeSigma Supplier No. ABE532

Rabbit Polyclonal Antibody

Anti-acetyl Histone H4 (Lys12) Antibody is a highly specific rabbit polyclonal antibody, that targets Histone H4 and has been tested in western blotting and Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl Histone H4 (Lys12)
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.0002% thimerosal.
Gene Symbols HHF1; HHF2
Host Species Rabbit
Immunogen Ovalbumin-conjugated linear peptide corresponding to yeast Histone H4 acetylated at Lys12.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_009563
Target Species Human, Yeast
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.