Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-acetyl-Histone H4 (Lys16), Alexa Fluor 488, Polyclonal

Catalog No. 07329AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07-329-AF488 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-329-AF488 Supplier MilliporeSigma™ Supplier No. 07329AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects acetyl-Histone H4 (Lys16) Clone: in Human, Mouse samples, and it is validated for Immunohistochemistry

Histone H4 (UniProt P02309) is encoded by the HHF1 (ORF YBR0122; Gene ID 852294) and HHF2 (ORF N2752; Gene ID 855701) genes in yeast species. Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H4 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H4 (Lys16)
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Purified rabbit antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide.
Gene Accession No. P02309
Gene Symbols HHF1, YBR009C, YBR0122, HHF2, YNL030W, N2752
Host Species Rabbit
Immunogen Ovalbumin-conjugated, synthetic peptide (…GAAcKRH …) in which AcK corresponds to residue 16 of yeast histone H4.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_009563.1
Test Specificity H4K16ac specificity of the unconjugated antibody has been confirmed using the Absurance™ Histone H3 Antibody Specificity Array and the Absurance™ Histone H2A, H2B, H4 Antibody Specificity Array.
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.