Learn More
MilliporeSigma™ anti-acetyl Histone H4 (Lys5/8/12/16), Clone: 3HH4-4C10, ™
Supplier: MilliporeSigma™ 051355/DEL

Description
Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the «beads on a string« structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs). Posttranslational acetylation of the conserved core histone N-terminal tail domains is typically associated with gene activation.
Host Species: Mouse
Clone: 3HH4-4C10
Isotype: IgG1, κ
Cross Reactivity: Human, Mouse, Rat
Immunogen: Linear peptide corresponding to human Histone H4 acetylated at Lys5/8/12/16.
Immunocytochemistry, Western Blotting

Specifications
acetyl Histone H4 (Lys5/8/12/16) | |
3HH4-4C10 | |
Mouse | |
100 μL | |
Human, Mouse, Rat | |
IgG1 κ |
Monoclonal | |
Unconjugated | |
Linear peptide corresponding to human Histone H4 acetylated at Lys5/8/12/16. | |
Primary | |
Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.